Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Detection Enhancer, for real-time PCR

Formulated to improve template amplification in real-time PCR (qPCR) workflows

$14.50 - $113.00

Specifications

Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method 1-step RT-qPCR
Reaction Speed Standard
For Use With (Equipment) 7500 System
View More Specs

Products 3
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
A44811
Encompass_Preferred
View Documents
Applied Biosystems™
A44811
1000 x 25 μL Reactions
Each for $113.00
Only null left
Edge
Add to Cart
 
A44941
Encompass_Preferred
View Documents
Applied Biosystems™
A44941
500 x 25 μL Reactions
Each for $68.00
Only null left
Edge
Add to Cart
 
A44810
Encompass_Preferred
View Documents
Applied Biosystems™
A44810
100 x 25 μL Reactions
Each for $14.50
Only null left
Add to Cart
 
Description

Description

  • Detection Enhancer is uniquely formulated to improve template amplification in real-time PCR (qPCR) workflows. This reagent is particularly beneficial with DNA samples containing GC-rich sequences and/or persistent secondary structure. Detection Enhancer is a component of the AgPath-ID One-Step RT-qPCR Reagents kit and is offered here as a stand-alone product that can be used with any Applied Biosystem real-time PCR master mix.
  • Improve qPCR amplification of templates containing high GC content and persistent secondary structure
  • Compatible with any Applied Biosystem real-time PCR master mix
  • Flexible kit sizes for a range of laboratory throughputs
  • GC-rich DNA sequences are usually defined as those that contain >60% cytosine (C) or guanine (G). GC-rich samples can be particularly challenging to amplify because the sequences are more stable than those with lower GC content. In qPCR experiments, this results in a higher DNA melting point. In addition, GC-rich sequences can form secondary structure, which can add further amplification challenges. Adding a small volume of Detection Enhancer to your reaction mix can dramatically improve qPCR amplification in GC-rich samples.
  • Detection Enhancer is specifically formulated to address qPCR amplification in particularly stable DNA sequences. It is only for use in real-time PCR workflows where amplification is affected by templates that are GC rich and/or contain persistent secondary structure. The reagents may affect sensitivity for other target types, so it is recommended to first assemble the reaction without Detection Enhancer. If no signal is observed from samples that are expected to be positive, then add Detection Enhancer to the reaction.
  • The recommended volume of Detection Enhancer per 25 μL reaction is 1.67 μL. For more information on applications of Detection Enhancer, please see the AgPath-ID One-Step RT-qPCR Reagents kit user guide
Specifications

Specifications

Primer-probe
1-step RT-qPCR
7500 System
Detection Enhancer
Real Time PCR (qPCR)
High
Standard
Applied Biosystems™
Dry Ice
Viral Samples
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.