Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Devazepide

Catalog No. 230410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-041-0 10 mg
23-045-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-041-0 Supplier Tocris Bioscience™ Supplier No. 2304/10
This item is not returnable. View return policy

Selective, orally active CCK1 receptor antagonist

Potent, orally active CCK1 (CCK-A) receptor antagonist that displays appetite-stimulant effects. Blocks the anorectic response to CCK-8 and increases food intake in rats following systemic and i.c.v administration.

Chemical Identifiers

CAS 103420-77-5
Molecular Formula C25H20N4O2

Specifications

Quantity 10 mg
Formula Weight 408.46
Purity >99%
Molecular Weight (g/mol) 408.461
Chemical Name or Material Devazepide
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.