Learn More
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Pump Sensor Cartridge, North America
Catalog No. 170000070 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID
Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
Hydrogen Sulfide, LEL-I
Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-000-0070 Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
17-000-0001 Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID
17-000-0071 Hydrogen Sulfide, LEL-I
17-000-0073 Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-000-0070 Supplier Blackline Safety™ Supplier No. G7CPHIMONA
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Wireless connectivity ensures that each G7c is remotely configurable and firmware updates are automatic. G7c supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving
Specifications
Components Measured Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
Type Multigas Cartridge