Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dexamethasone, MP Biomedicals™

Catalog No. p-4679726 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 mg
5 g

Anti-inflammatory agent

Dexamethasone is a glucocorticoid anti-inflammatory agent. It regulates T cell survival, growth, and differentiation. It also inhibits the induction of nitric oxide synthase.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dexamethasone
CAS 50-02-2
Synonym maxidex
Quantity 500 mg
Molecular Formula C22H29FO5
Formula Weight 392.47
Physical Form Powder
Color White
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.