Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Dexmedetomidine hydrochloride
GREENER_CHOICESDP

Catalog No. 274950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-495-0 50 mg
27-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-495-0 Supplier R&D Systems™ Supplier No. 2749/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, highly selective α2 agonist. Active isomer of medetomidine (Cat. No. 5160)

Dexmedetomidine hydrochloride is an active isomer of medetomidine, a potent, highly selective α2-adrenoceptor agonist (Ki values are 1.08 and 1750 nM for α2- and α1-adrenoceptors respectively). Displays greater selectivity over α1-adrenoceptors than Clonidine (Cat. No. 0690) and UK 14,304 (1620-, 220- and 300-fold respectively). Active in vivo; displays hypotensive, bradycardic, sedative, anxiolytic, hypothermic and analgesic effects.

Racemate also available.

Chemical Identifiers

CAS 145108-58-3
Molecular Formula C13H17ClN2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 236.74
Purity 98%
Molecular Weight (g/mol) 236.743
Chemical Name or Material Dexmedetomidine hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.