Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dexrazoxane hydrochloride
GREENER_CHOICE

Catalog No. 385710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-571-0 10 mg
38-575-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-571-0 Supplier Tocris Bioscience™ Supplier No. 3857/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Topoisomerase II inhibitor

Topoisomerase II inhibitor and intracellular ion chelator. Bridges and stabilizes an interface between two ATPase promoters to inhibit topoisomerase II activity. Cardioprotective when co-administered with doxorubicin; decreases formation of reactive oxygen species (ROS) and activates the PI3K/Akt survival pathway.

Chemical Identifiers

CAS 1263283-43-7
Molecular Formula C11H17ClN4O4

Specifications

Quantity 10 mg
Formula Weight 304.73
Purity >98%
Molecular Weight (g/mol) 304.731
Chemical Name or Material Dexrazoxane hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.