Learn More
Specifications
Specifications
| Melting Point | 83°C |
| Color | White |
| Density | 1.54g/mL |
| Packaging | Poly Pail |
| Linear Formula | CH2OHCHO(CHOH)3CHOH·H2O |
| Quantity | 3 kg |
| Residual Solvents | Meets Requirements (USP) |
| Vapor Pressure | Negligible |
| Formula Weight | 198.17 |
| Grade | USP |
| Show More |
Safety and Handling
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.