Learn More
Description
- Exclusive design that minimizes the pressure points and fits comfortably on most faces
- Humidity and moisture resistant filter media that repels moisture for a longer lasting respirator
- Strong, rigid outer shell prevents masks from collapsing in high humidity
- Dual welded head straps secure the respirator comfortably
- Latex free woven head straps reduce the possibility of breakage
Specifications
Specifications
| Aerosol Type | N - Not Resistant to Oil |
| Certifications (NIOSH) | NIOSH 42 CFR 84, N95 |
| Design Type | Flat-fold |
| Disposable | Single-use |
| Filter Type | N95 |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| With Exhalation Valve | No |
| Filter Efficiency | 95% |
| Latex Free | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.