Learn More
DFF40/CAD Antibody (36A349) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody has been used in 3 publications
Supplier:  Novus Biologicals NB12013592SS
 View more versions of this product
Catalog No. NB12013592T
Only null left
Add to Cart
Description
Description
DFF40/CAD Monoclonal specifically detects DFF40/CAD in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
DFF40/CAD
Monoclonal
0.5 mg/mL
Western Blot 1:100-1:2000
DFFB
Anti-DFF40 monoclonal antibody was raised against a peptide corresponding to amino acids 205 to 222 of human DFF40.
0.025 mg
Primary
In HeLa, a 40 kDa band is observed.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
36A349
Unconjugated
CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.-
Mouse
Protein G purified
RUO
1677
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only