Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFF40/CAD Antibody (36A349) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$206.50 - $426.00

Specifications

Antigen DFF40/CAD
Clone 36A349
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB12013592T
SDP
View Documents
Novus Biologicals
NB12013592SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB12013592
SDP
View Documents
Novus Biologicals
NB12013592
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

DFF40/CAD Monoclonal specifically detects DFF40/CAD in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

DFF40/CAD
0.5 mg/mL
Western Blot
Unconjugated
Mouse
Human
1677
Anti-DFF40 monoclonal antibody was raised against a peptide corresponding to amino acids 205 to 222 of human DFF40.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
36A349
Western Blot 1:100-1:2000
Monoclonal
Purified
RUO
CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.-
DFFB
IgG
Protein G purified
In HeLa, a 40 kDa band is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.