Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFF40/CAD Antibody (36A349), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB338931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338931 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338931 Supplier Novus Biologicals Supplier No. NB12013592JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DFF40/CAD Monoclonal antibody specifically detects DFF40/CAD in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen DFF40/CAD
Applications Western Blot
Classification Monoclonal
Clone 36A349
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.-
Host Species Mouse
Immunogen Anti-DFF40 monoclonal antibody was raised against a peptide corresponding to amino acids 205 to 222 of human DFF40.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 1677
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.