Learn More
Description
Specifications
Specifications
| Antigen | DFF40/CAD |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 ug/mL |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | NP_031885 |
| Gene Alias | CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- |
| Gene Symbols | DFFB |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
