Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFF40/CAD Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB356981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356981 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356981 Supplier Novus Biologicals Supplier No. NBP176948MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DFF40/CAD Polyclonal antibody specifically detects DFF40/CAD in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen DFF40/CAD
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.-
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 18 amino acids near the center of murine CAD. The immunogen is located within amino acids 130 - 180 of DFF40. Amino Acid Squence: GLESRFRNKSGYLRYSC
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 1677
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.