Learn More
Description
Specifications
Specifications
| Antigen | DFF45/ICAD |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | O54786 |
| Gene Alias | DFF1DNA fragmentation factor 45 kDa subunit, DFF45DNA fragmentation factor subunit alpha, DFF-45DNA fragmentation factor, 45 kD, alpha polypeptide, DNA fragmentation factor, 45kDa, alpha polypeptide, ICAD, Inhibitor of CAD |
| Gene Symbols | DFFA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
