Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DFMO
GREENER_CHOICESDP

Catalog No. 276110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-611-0 10 mg
27-615-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-611-0 Supplier Tocris Bioscience™ Supplier No. 2761/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ornithine decarboxylase inhibitor

Irreversible inhibitor of ornithine decarboxylase (ODC) that inhibits polyamine biosynthesis. Displays antiapoptotic, antiangiogenic and antiparasitic activity. Induces re-expression of aberrantly silenced tumor suppressor genes when used in combination with oligoamine analogs.

Chemical Identifiers

CAS 68278-23-9
Molecular Formula C6H13ClF2N2O2

Specifications

Quantity 10 mg
Formula Weight 218.63
Purity >98%
Molecular Weight (g/mol) 218.629
Chemical Name or Material DFMO
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.