Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGAT1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB11041487B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11041487B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11041487B Supplier Novus Biologicals Supplier No. NB11041487B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DGAT1 Polyclonal specifically detects DGAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting.

Specifications

Antigen DGAT1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O75907
Gene Alias ACAT-related gene product 1, AGRP1, ARGP1acyl coenzyme A:cholesterol acyltransferase related gene 1, DGATACAT related gene product 1, diacylglycerol O-acyltransferase 1, diacylglycerol O-acyltransferase homolog 1, diacylglycerol O-acyltransferase homolog 1 (mouse), Diglyceride acyltransferase, EC 2.3.1, EC 2.3.1.20
Gene Symbols DGAT1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 200-300) of human DGAT1. [Swiss-Prot# O75907]
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8694
Test Specificity Human, mouse, bovine, Zebrafish, and rat. Predicted to react with primate, sheep, and goat based on 100% sequence homology.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.