Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103785 Supplier Invitrogen™ Supplier No. PA5103785
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DGAT2.

Acyl-coenzyme A:diacylglycerol acyltransferase, also known as DGAT1 and ARGP1, is a microsomal enzyme that assists in the synthesis of fatty acids into triglycerides. DGAT1 catalyzes the terminal and only committed step in triacylglycerol synthesis by using diacylglycerol (DAG) and fatty acyl CoA as substrates. DGAT1 plays a fundamental role in the metabolism of cellular diacylglycerol and is important in higher eukaryotes for physiologic processes involving triacylglycerol metabolism, such as intestinal fat absorption, lipoprotein assembly, adipose tissue form-ation and lactation. DGAT2, which has no homology to DGAT1, differs from DGAT1 in that its activity has been shown to be inhibited by MgCl in an in vitro assay. DGAT2 is expressed primarily in liver and white adipose tissue, which suggests that it plays an important role in mammalian triglyceride metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGAT2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DGAT2
Gene Accession No. Q5FVP8, Q96PD7, Q9DCV3
Gene Alias 0610010B06Rik; Acyl-CoA retinol O-fatty-acyltransferase; ARAT; DGAT2; DGAT-2; diacylglycerol acyltransferase 2; diacylglycerol O-acyltransferase 2; diacylglycerol O-acyltransferase homolog 2; diacylglycerol O-acyltransferase-like protein 2; diglyceride acyltransferase 2; DKFZp686A15125; GS1999FULL; HMFN1045; LOW QUALITY PROTEIN: diacylglycerol O-acyltransferase 2; putative diacylglycerol O-acyltransferase; retinol O-fatty-acyltransferase; UNQ738/PRO1433
Gene Symbols DGAT2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DGAT2(Accession Q96PD7), corresponding to amino acid residues N155-R205.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252900, 67800, 84649
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.