Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGCR14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP236530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236530 Supplier Novus Biologicals Supplier No. NBP236530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DGCR14 Polyclonal specifically detects DGCR14 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen DGCR14
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2500, Immunoprecipitation 2 - 10 ug/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_073210.1
Gene Alias DGCR13, DGS-HDGS-I, DGSIDGSH, DiGeorge syndrome critical region 13, DiGeorge syndrome critical region 14, DiGeorge syndrome critical region gene 13, DiGeorge syndrome critical region gene 14, DiGeorge syndrome gene H, DiGeorge syndrome gene I, DiGeorge syndrome protein H, ES2DiGeorge syndrome critical region gene DGSI, Es2el, Protein DGCR13, protein DGCR14, Protein ES2
Gene Symbols ESS2
Host Species Rabbit
Immunogen The immunogen recognized by the antibody maps to a region between residue 426 to 476 of human DiGeorge Syndrome Critical Region Gene 14 using the numbering given in entry NP_073210.1.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8220
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.