Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGCR8 Rabbit anti-Human, Mouse, Clone: 3T5Q6, Novus Biologicals™
SDP

Catalog No. NB167704 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167704 20 μg
NB167703 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167704 Supplier Novus Biologicals Supplier No. NBP31630620UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DGCR8 Monoclonal antibody specifically detects DGCR8 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen DGCR8
Applications Western Blot
Classification Monoclonal
Clone 3T5Q6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias C22orf12, DGCRK6chromosome 22 open reading frame 12, DiGeorge syndrome critical region 8, DiGeorge syndrome critical region gene 8, Gy1, microprocessor complex subunit DGCR8, pasha
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 436-764 of human DGCR8 (Q8WYQ5).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Epigenetics, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54487
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.