Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGK-alpha Antibody (OTI4A11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N270547A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270547A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270547A700 Supplier Novus Biologicals Supplier No. NBP270547AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DGK-alpha Monoclonal specifically detects DGK-alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen DGK-alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4A11
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DAG kinase alpha, DAGK, DAGK1diacylglycerol kinase, alpha (80kD), DGK-alphaEC 2.7.1.107, diacylglycerol kinase alpha, diacylglycerol kinase, alpha 80kDa, Diglyceride kinase alpha, MGC12821,80 kDa diacylglycerol kinase, MGC42356
Gene Symbols DGKA
Host Species Mouse
Immunogen Full length recombinant protein of human DGK-alpha (NP_001336) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1606.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.