Learn More
Description
Specifications
Specifications
| Antigen | DGK-alpha |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI4A11 |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | DAG kinase alpha, DAGK, DAGK1diacylglycerol kinase, alpha (80kD), DGK-alphaEC 2.7.1.107, diacylglycerol kinase alpha, diacylglycerol kinase, alpha 80kDa, Diglyceride kinase alpha, MGC12821,80 kDa diacylglycerol kinase, MGC42356 |
| Gene Symbols | DGKA |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
