Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGK-beta Antibody (OTI1F9), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270552V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270552V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270552V Supplier Novus Biologicals Supplier No. NBP270552V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DGK-beta Monoclonal specifically detects DGK-beta in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot.

Specifications

Antigen DGK-beta
Applications Western Blot
Classification Monoclonal
Clone OTI1F9
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGK-beta, diacylglycerol kinase beta, diacylglycerol kinase, beta (90kD), diacylglycerol kinase, beta 90kDa, Diglyceride kinase beta, EC 2.7.1, EC 2.7.1.107, KIAA0718DGKDGK-BETA
Gene Symbols DGKB
Host Species Mouse
Immunogen Full length recombinant protein of human DGK-beta (NP_663733) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1607.0
Target Species Human, Mouse, Rat, Canine, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.