Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGK-epsilon Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | DGK-epsilon |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DGK-epsilon Polyclonal specifically detects DGK-epsilon in Human, Mouse samples. It is validated for Western Blot.Specifications
| DGK-epsilon | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| DAG kinase epsilon, DAGK5, DAGK6, DGK, DGK-epsilon, diacylglycerol kinase epsilon, diacylglycerol kinase, epsilon (64kD), diacylglycerol kinase, epsilon 64kDa, Diglyceride kinase epsilon, EC 2.7.1.107 | |
| DGKE | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P52429 | |
| 8526 | |
| Synthetic peptides corresponding to DGKE(diacylglycerol kinase, epsilon 64kDa) The peptide sequence was selected from the N terminal of DGKE. Peptide sequence EAERRPAPGSPSEGLFADGHLILWTLCSVLLPVFITFWCSLQRSRRQLHR. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title