Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGK-zeta Antibody (RB3716), Novus Biologicals™
SDP

Catalog No. NBP229920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229920 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229920 Supplier Novus Biologicals Supplier No. NBP229920

Rabbit Polyclonal Antibody

DGK-zeta Polyclonal specifically detects DGK-zeta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DGK-zeta
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone RB3716
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:50, Immunohistochemistry-Paraffin 1:50 - 1:100
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. Q13574
Gene Alias DAGK5, DAGK6DAG kinase zeta, DGK-ZETA, diacylglycerol kinase zeta, diacylglycerol kinase, zeta, diacylglycerol kinase, zeta 104kDa, Diglyceride kinase zeta, EC 2.7.1.107, hDGKzeta
Gene Symbols DGKZ
Host Species Rabbit
Immunogen This DGKZ antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 989-1019 amino acids from the C-terminal region of human DGKZ.
Molecular Weight of Antigen 124.128 kDa
Purification Method Protein A purified
Quantity 0.4 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8525
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.