Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DH5α Competent Cells, High Efficiency PROMO
Click to view available options
Quantity:
20 Reactions
Description
- The φ80dlacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors to allow blue/white colony screening on bacterial agar plates containing Bluo-Gal or X-Gal
- RecA1 and endA1 mutations in DH5α cells improve insert stability and quality of the extracted plasmid DNA
- High transformation efficiency: >1x109 cfu/μg pUC19 DNA
- Suitable for routine and high-throughput cloning applications
- Genetic markers that allow for blue/white colony screening
- Convenient two reactions per vial packaging
- S.O.C. Outgrowth medium included
- DH5α Competent Cells are suitable for variety of applications requiring high transformation efficiency:
- Efficient DNA cloning derived from PCR, cDNA-generation reactions
- recA1 marker facilitates working with difficult to transform DNA
- Ideal for generation of cDNA libraries using plasmid-derived vectors
- Site-directed mutagenesis
- Genotype: F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1
Specifications
Specifications
Description | Chemically Competent |
Contains F' Episome | Lacks F' Episome |
Content And Storage | Dry Ice (-80°C) |
Bacterial or Yeast Strain | DH5α |
Species | E. coli |
Format | Tube |
For Use With (Application) | Cloning |
Quantity | 20 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction