Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHPS Antibody (OTI2C9), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N270570A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270570A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270570A405 Supplier Novus Biologicals Supplier No. NBP270570AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DHPS Monoclonal specifically detects DHPS in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen DHPS
Applications Western Blot
Classification Monoclonal
Clone OTI2C9
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias deoxyhypusine synthase, DHS, DS, EC 2.5.1, EC 2.5.1.46, hypusine, MIG13, migration-inducing gene 13
Gene Symbols DHPS
Host Species Mouse
Immunogen Full length recombinant protein of human DHPS (NP_001921) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1725.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.