Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS3 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB180747 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180747 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180747 Supplier Novus Biologicals Supplier No. NBP306444AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DHRS3 Polyclonal antibody specifically detects DHRS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen DHRS3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias DD83.1, dehydrogenase/reductase (SDR family) member 3, EC 1.1.1, EC 1.1.1.300, RDH17, Retinal short-chain dehydrogenase/reductase 1, RETSDR1, retSDR1short chain dehydrogenase/reductase family 16C, member 1, Rsdr1, SDR1, SDR16C1, short-chain dehydrogenase/reductase 1, short-chain dehydrogenase/reductase 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DHRS3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 9249
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.