Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS4 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB054691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054691 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054691 Supplier Novus Biologicals Supplier No. NBP298675F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHRS4 Polyclonal specifically detects DHRS4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DHRS4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CR, dehydrogenase/reductase (SDR family) member 4, dehydrogenase/reductase (SDR family) member 4 like 2A, dehydrogenase/reductase SDR family member 4, EC 1.1.1, EC 1.1.1.184, FLJ11008, humNRDR, NADP-dependent retinol dehydrogenase, NRDRNADPH-dependent retinol dehydrogenase/reductase, Peroxisomal short-chain alcohol dehydrogenase, PHCR, PSCD, SCAD-SRLNADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase, SDR25C1, SDR25C2, SDR-SRL, short chain dehydrogenase/reductase family 25C, member 1, short chain dehydrogenase/reductase family 25C, member 2, Short-chain dehydrogenase/reductase family member 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DHRS4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10901
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.