Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX30 Antibody, Novus Biologicals™
SDP

Catalog No. NBP126604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126604 Supplier Novus Biologicals Supplier No. NBP126604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHX30 Polyclonal specifically detects DHX30 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen DHX30
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 5-10 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q7L2E3
Gene Alias ATP-dependent RNA helicase DHX30, DDX30, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30, DEAH (Asp-Glu-Ala-His) box polypeptide 30, DEAH box protein 30, EC 3.6.1, EC 3.6.4.13, FLJ11214, KIAA0890RETCOR, putative ATP-dependent RNA helicase DHX30, retina co-repressor
Gene Symbols DHX30
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1144 and 1194 of human DEAH (Asp-Glu-Ala-His) box polypeptide 30 using the numbering given in entry NP_619520.1 (GeneID 22907).
Molecular Weight of Antigen 134 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22907
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.