Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX33 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292296 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292296 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292296 Supplier Novus Biologicals Supplier No. NB1002581PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHX33 Polyclonal specifically detects DHX33 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen DHX33
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias DDX33, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 33, DEAH (Asp-Glu-Ala-His) box polypeptide 33, DEAH box protein 33, DKFZp762F2011, EC 3.6.1, EC 3.6.4.13, FLJ21972, putative ATP-dependent RNA helicase DHX33
Host Species Rabbit
Immunogen Maps to a region between residue 650 and the C-terminus (residue 707) of human DEAH (Asp-Glu-Ala-His) Box Polypeptide 33 using the numbering given in entry NP_064547.2 (GeneID 56919).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56919
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.