Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX34 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15729420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15729420 20 μL
NBP157294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15729420 Supplier Novus Biologicals Supplier No. NBP15729420UL

Rabbit Polyclonal Antibody

DHX34 Polyclonal specifically detects DHX34 in Human samples. It is validated for Western Blot.

Specifications

Antigen DHX34
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Alias DDX34, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 34, DEAH (Asp-Glu-Ala-His) box polypeptide 34, DEAH box protein 34, HRH1, KIAA0134
Gene Symbols DHX34
Host Species Rabbit
Immunogen Synthetic peptides corresponding to DHX34(DEAH (Asp-Glu-Ala-His) box polypeptide 34) The peptide sequence was selected from the middle region of DHX34. Peptide sequence VPGRLFPITVVYQPQEAEPTTSKSEKLDPRPFLRVLESIDHKYPPEERGD.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9704
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.