Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX37 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB335268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335268 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335268 Supplier Novus Biologicals Supplier No. NBP318056MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHX37 Polyclonal antibody specifically detects DHX37 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen DHX37
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DDX37, DEAD/DEAH box helicase DDX37, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 37, DEAH (Asp-Glu-Ala-His) box polypeptide 37, DEAH box protein 37, EC 3.6.1, EC 3.6.4.13, FLJ41974, KIAA1517MGC46245, MGC2695, MGC4322, probable ATP-dependent RNA helicase DHX37
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human DHX37 (between amino acids 650-700) [UniProt Q8IY37]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 57647
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.