Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX57 Antibody, Novus Biologicals™
SDP

Catalog No. NBP128693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128693 Supplier Novus Biologicals Supplier No. NBP128693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHX57 Polyclonal specifically detects DHX57 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen DHX57
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 10 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_945314.1
Gene Alias DDX57, DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57, DEAH box protein 57, DEAH-box RNA/DNA helicase AAM73547, EC 3.6.1, EC 3.6.4.13, FLJ32861, putative ATP-dependent RNA helicase DHX57
Gene Symbols DHX57
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57 using the numbering given in entry NP_945314.1 (GeneID 90957).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90957
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.