Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX58 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB358268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358268 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358268 Supplier Novus Biologicals Supplier No. NBP176389MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHX58 Polyclonal antibody specifically detects DHX58 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen DHX58
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias D11lgp2e, D11LGP2probable ATP-dependent RNA helicase DHX58, DEXH (Asp-Glu-X-His) box polypeptide 58, EC 3.6.4.13, LGP2ortholog of mouse D11lgp2, Probable ATP-dependent helicase LGP2, Protein D11Lgp2 homolog, RIG-I-like receptor LGP2, RLR, RNA helicase LGP2
Host Species Rabbit
Immunogen Antibody was raised against a 11 amino acid synthetic peptide from near the center of human LGP2. The immunogen is located within amino acids 200 - 250 of LGP2. Amino Acid Squence: HRRSQDPFGDL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 79132
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.