Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX8 Rabbit anti-Human, Clone: S04-1I7, Novus Biologicals™
SDP

Catalog No. NB188399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188399 25 μg
NB188398 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188399 Supplier Novus Biologicals Supplier No. NBP31502325UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DHX8 Monoclonal antibody specifically detects DHX8 in Human samples. It is validated for Western Blot

Specifications

Antigen DHX8
Applications Western Blot
Classification Monoclonal
Clone S04-1I7
Conjugate Unconjugated
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias DDX8ATP-dependent RNA helicase DHX8, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 8 (RNA helicase), DEAH (Asp-Glu-Ala-His) box polypeptide 8, DEAH box protein 8, DEAH-box protein 8, EC 3.6.1, EC 3.6.4.13, HRH1RNA helicase HRH1, PRP22, PRPF22
Host Species Rabbit
Immunogen A synthetic peptide of human DHX8 (Uniprot#: Q14562)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1659
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.