Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX9 Monoclonal Antibody (8E3), Active Motif™
SDP

Rat Monoclonal Antibody

$159.90 - $633.45

Specifications

Antigen DHX9
Clone 8E3
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3634
SDP
Encompass
View Documents
Active Motif
61706
10 μg
Each for $159.90
Only null left
Add to Cart
 
50-199-3639
SDP
Encompass
View Documents
Active Motif
61705
100 μg
Each for $633.45
Only null left
Add to Cart
 
Description

Description

The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.
Specifications

Specifications

DHX9
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
AI326842; ATP-dependent RNA helicase A; BOS_16077; Ddx9; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; Dhx9; FLJ17406; HEL-5; Leukophysin; LKP; mHEL-5; NDH II; NDH2; NDHII; Nuclear DNA helicase II; OTTHUMP00000033287; RHA; RNA helicase; RNA helicase A
Dhx9
IgG1
Protein A
Antibody
8E3
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rat
Human, Mouse
O70133, Q08211
13211, 1660
This antibody was raised against a peptide within the N-terminal region of mouse DHX9.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Dhx9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.