Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX9 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$167.95

Specifications

Antigen DHX9
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-155-8626
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A300855AT
20 μL
Each of 1 for $167.95
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

This gene encodes a member of the DEAH-containing family of RNA helicases. The encoded protein is an enzyme that catalyzes the ATP-dependent unwinding of double-stranded RNA and DNA-RNA complexes. This protein localizes to both the nucleus and the cytoplasm and functions as a transcriptional regulator. This protein may also be involved in the expression and nuclear export of retroviral RNAs. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 11 and 13.
Specifications

Specifications

DHX9
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
DHX9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DDX9; LKP; NDHII; RHA; ATP-dependent RNA helicase A; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNA helicase II; leukophysin); DEAD/H box-9 (nuclear DNA helicase II; RNA helicase A); OTTHUMP00000033287; leukophysin
DHX9
IgG
Antigen affinity chromatography
Antibody
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human
Q08211
1660
Between 1220 and C-term
Primary
4° C
DHX9
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.