Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butyl Ether, +99%, Extra Pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Metal drum
142-96-1
C8H18O
130.23
MFCD00009461
DURPTKYDGMDSBL-UHFFFAOYSA-N
di-n-butyl ether,butyl ether,dibutyl ether,butane, 1,1'-oxybis,n-butyl ether,dibutyl oxide,butyl oxide,ether butylique,1,1'-oxybis butane,di-n-butylether
8909
1-butoxybutane
CCCCOCCCC
142-96-1
C8H18O
130.23
MFCD00009461
DURPTKYDGMDSBL-UHFFFAOYSA-N
di-n-butyl ether,butyl ether,dibutyl ether,butane, 1,1'-oxybis,n-butyl ether,dibutyl oxide,butyl oxide,ether butylique,1,1'-oxybis butane,di-n-butylether
8909
1-butoxybutane
CCCCOCCCC
Viscosity 0.69 mPa.s (20°C)
Specific Gravity 0.768
Boiling Point 141.0°C
Water 0.05% max.
Refractive Index 1.3978 to 1.3998
Melting Point -95.0°C
Packaging Glass Bottle
Infrared Spectrum Authentic
Density 0.7680g/mL
Quantity 1L
Color Undesignated
Flash Point 25°C
Assay Percent Range 99+%
Linear Formula [CH3(CH2)3]2O
Beilstein 01, 369
Merck Index 15, 1572
Solubility Information Solubility in water: 0.03% (20°C). Other solubilities: miscible with ethanol,ether,acetone
Formula Weight 130.23
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Di-n-butyl ether
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning