Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Di-tert-butyl L-tartrate, 99%

Catalog No. AAH5586003
Click to view available options
Quantity:
250 mg
500 mg
1 g

CAS: 117384-45-9 | C12H22O6 | 262.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117384-45-9
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.30
MDL Number MFCD00192000
InChI Key ITWOKJQQGHCDBL-UHFFFAOYNA-N
Synonym +-di-tert-butyl l-tartrate, di-tert-butyl l-+-tartrate, 2r,3r-di-tert-butyl 2,3-dihydroxysuccinate, di-t-butyl l-tartrate, r,r-di-tert-butyl-tartrat, di-tert-butyll-+-tartrate, l-tartaric acid di-tert-butyl ester, l-+-tartaric acid di-tert-butyl ester, 1,4-di-tert-butyl 2r,3r-2,3-dihydroxybutanedioate
PubChem CID 9943166
IUPAC Name ditert-butyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES CC(C)(C)OC(=O)C(O)C(O)C(=O)OC(C)(C)C

Specifications

Melting Point 90°C to 92°C
Color White
Density 1.143 g/mL
Flash Point 109°C (228°F)
Quantity 1 g
Beilstein 5026723
Formula Weight 262.3
Percent Purity 99%
Physical Form Solid
Chemical Name or Material (+)-Di-tert-butyl L-tartrate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.