Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Diamode Antidiarrheal Caplets

Catalog No. p-7078228 Shop All Medique Products
Click to view available options
Quantity:
2/Pk., 100/Box

For control and symptomatic relief of acute non-specific diarrhea.

  • Easy to swallow
  • Film coated
  • Each caplet contains: Lopermide HCl 2mg

Specifications

Product Type Anti-diarrheal Caplets
Active Ingredients Lopermide HCl (2 mg)
Format Film-coated Caplet
Color Green
For Use With (Application) For control and symptomatic relief of acute non-specific diarrhea
Quantity 2/Pk., 100/Box
Recommended Age 12 years and older
Storage Requirements 68°F to 77°F (20°C to 25°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.