Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIAPH-2 Antibody (V78P3C10*D3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250229FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250229FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250229FR Supplier Novus Biologicals Supplier No. NBP250229FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DIAPH-2 Monoclonal specifically detects DIAPH-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen DIAPH-2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V78P3C10*D3
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DIA2, DIAdiaphanous (Drosophila, homolog) 2, diaphanous homolog 2 (Drosophila), Diaphanous-related formin-2, diaphorase-2, DRF2, POF, POF2FLJ11167, protein diaphanous homolog 2
Gene Symbols DIAPH2
Host Species Mouse
Immunogen Peptide Sequence - RHNGAISSK
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1730
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.