Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIAPH1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB600230H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600230H 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600230H Supplier Novus Biologicals Supplier No. NB600230H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DIAPH1 Polyclonal specifically detects DIAPH1 in Human samples. It is validated for Western Blot, Knockdown Validated.

Specifications

Antigen DIAPH1
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, KnockDown Validated
Formulation PBS with No Preservative
Gene Alias DIA1, diaphanous homolog 1 (Drosophila), hDIA1, homolog) 1, LFHL1FLJ25265
Gene Symbols DIAPH1
Host Species Rabbit
Immunogen A synthetic peptide, which represented a portion of human diaphanous homolog 1 encoded within exon 1 (LocusLink ID 1729).
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1729
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.