Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diatomaceous Earth, acid-washed, ∽95% SiO2, MP Biomedicals™

Catalog No. p-4405337 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 kg
5 kg

Chemical Identifiers

CAS 61790-53-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Diatomaceous silica
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form Powder
Quantity 1 kg
Color Pink to Tan
Assay Percent Range ∽95%
Formula Weight 60.04
Chemical Name or Material Diatomaceous Earth, Non-washed
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.