Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzylamine, 98%

Catalog No. p-7081500
Click to view available options
Quantity:
250 g
1000 g

CAS: 103-49-1 | C14H15N | 197.28 g/mol

Dibenzylamine is used to prepare rubber accelerator for the vulcanization process and reaction-stoppers. Dibenzylamine type accelerators reduce nitrosamine production in the compounding process. It is also used to produce dibenzyl-nitroso-amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzylamine is used to prepare rubber accelerator for the vulcanization process and reaction-stoppers. Dibenzylamine type accelerators reduce nitrosamine production in the compounding process. It is also used to produce dibenzyl-nitroso-amine.

Solubility
Soluble in water(0.4g/L).

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-49-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00004770
InChI Key BWLUMTFWVZZZND-UHFFFAOYSA-N
Synonym dibenzylamine, benzenemethanamine, n-phenylmethyl, n-benzylbenzylamine, bibenzylamine, dibenzyl-amine, dibenzyl amine, bisbenzylamine, n,n-dibenzylamine, n-benzylaminomethyl benzene, unii-3g0yfx01c6
PubChem CID 7656
SMILES C(NCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -26°C
Density 1.026
pH 8.9
Boiling Point 300°C
Flash Point 143°C (289°F)
Odor Weak
Refractive Index 1.5745
Quantity 250 g
UN Number UN2735
Beilstein 909664
Merck Index 14,3011
Solubility Information Soluble in water(0.4g/L).
IUPAC Name N-benzyl-1-phenylmethanamine
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material Dibenzylamine
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 203-117-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.