Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dibutyryl-cAMP, sodium salt
GREENER_CHOICE

Catalog No. p-4783408
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cell-permeable cAMP analog

Cell-permeable analog of cAMP; activates cAMP-dependent protein kinases. Promotes differentiation of dopaminergic neurons (iDA) from hPSCs in combination with other reagents.

Chemical Identifiers

CAS 16980-89-5
Molecular Formula C18H26N5NaO8P

Specifications

Quantity 10 mg
Formula Weight 491.37
Purity >98%
Molecular Weight (g/mol) 494.397
Chemical Name or Material Dibutyryl-cAMP, sodium salt
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.