Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicer Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP106521FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106521FR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106521FR Supplier Novus Biologicals Supplier No. NBP106521FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dicer Polyclonal specifically detects Dicer in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Dicer
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DCR1, Dicer, dicer 1, double-stranded RNA-specific endoribonuclease, dicer 1, ribonuclease type III, Dicer1, Dcr-1 homolog (Drosophila), EC 3.1.26, EC 3.1.26.-, EC 3.1.26.3, Helicase MOI, Helicase with RNase motif, helicase-moi, HERNADicer1, Dcr-1 homolog, K12H4.8-LIKE, KIAA0928endoribonuclease Dicer
Gene Symbols DICER1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1175 and 1225 of human double-stranded RNA-specific endoribonuclease; dicer1 using the numbering given in entry NP_085124.2 (GeneID 23405).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication, Chromatin Research, Cytokine Research, Epigenetics, Immunology, Innate Immunity, microRNA, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23405
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.