Learn More
Description
Specifications
Specifications
| Antigen | Dicer |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:100 - 1:1000, ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) reported in scientific literature (PMID 16823371) |
| Formulation | Crude whole Rabbit sera, 50% Glycerol with 0.025% Sodium Azide |
| Gene Accession No. | Q9UPY3 |
| Gene Alias | DCR1, Dicer, dicer 1, double-stranded RNA-specific endoribonuclease, dicer 1, ribonuclease type III, Dicer1, Dcr-1 homolog (Drosophila), EC 3.1.26, EC 3.1.26.-, EC 3.1.26.3, Helicase MOI, Helicase with RNase motif, helicase-moi, HERNADicer1, Dcr-1 homolog, K12H4.8-LIKE, KIAA0928endoribonuclease Dicer |
| Gene Symbols | DICER1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
