Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicer Antibody (S167-7), FITC, Novus Biologicals™
SDP

Catalog No. NBP222415F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222415F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222415F Supplier Novus Biologicals Supplier No. NBP222415F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dicer Monoclonal specifically detects Dicer in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dicer
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S167-7
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias DCR1, Dicer, dicer 1, double-stranded RNA-specific endoribonuclease, dicer 1, ribonuclease type III, Dicer1, Dcr-1 homolog (Drosophila), EC 3.1.26, EC 3.1.26.-, EC 3.1.26.3, Helicase MOI, Helicase with RNase motif, helicase-moi, HERNADicer1, Dcr-1 homolog, K12H4.8-LIKE, KIAA0928endoribonuclease Dicer
Gene Symbols DICER1
Host Species Mouse
Immunogen Fusion protein amino acids 1638-1899 of mouse Endoribonuclease Dicer
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication, Chromatin Research, Cytokine Research, Epigenetics, Immunology, Innate Immunity, microRNA, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23405
Test Specificity Detects approx 215kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.