Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethyl Methyl Ether 97.0+%, TCI America™
SDP

Catalog No. D1645250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1645250G 250 g
D164525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1645250G Supplier TCI America Supplier No. D1645250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4885-02-3
Molecular Formula C2H4Cl2O
Molecular Weight (g/mol) 114.95
MDL Number MFCD00000828
InChI Key GRTGGSXWHGKRSB-UHFFFAOYSA-N
Synonym dichloromethyl methyl ether, 1,1-dichlorodimethyl ether, dichloro methoxy methane, methane, dichloromethoxy, ether, dichloromethyl methyl, alpha,alpha-dichloromethyl methyl ether, methoxydichloromethane, 1,1-dichloromethyl methyl ether, alpha,alpha-dichloromethyl ether, dichloromethylmethylether
PubChem CID 21004
IUPAC Name dichloro(methoxy)methane
SMILES COC(Cl)Cl

Specifications

Color Yellow
Boiling Point 85°C
Quantity 250 g
UN Number 2924
Formula Weight 114.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloromethyl Methyl Ether
RTECSNumber KN8150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.