Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethylenedimethyliminium Chloride 97.0+%, TCI America™
SDP

Catalog No. D159725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D159725G 25 g
D15975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D159725G Supplier TCI America Supplier No. D159725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33842-02-3
Molecular Formula C3H6Cl3N
Molecular Weight (g/mol) 162.44
MDL Number MFCD00011870
InChI Key NRNFKRFWZQQDMD-UHFFFAOYSA-M
Synonym dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
PubChem CID 2723945
IUPAC Name dichloro(dimethylamino)methylium chloride
SMILES [Cl-].CN(C)[C+](Cl)Cl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 162.44
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloromethylenedimethyliminium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.